accessibility menu, dialog, popup

UserName

IOH

CAS RN 29558-78-9

IUPAC Name: 4'-iodo-[1,1'-biphenyl]-4-ol
Synonyms: 4'-iodo-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 296.11
Molecular Formula: C12H9IO
InChi Key: WJXIAMCDWSUSEI-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(I)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results