accessibility menu, dialog, popup

UserName

CAS RN 294885-28-2

IUPAC Name: 1-tert-butyl 4-(2,3,4,5,6-pentafluorophenyl) piperidine-1,4-dicarboxylate
Synonyms: 1-tert-butyl 4-(2,3,4,5,6-pentafluorophenyl) piperidine-1,4-dicarboxylate
Molecular Weight (g/mol): 395.33
Molecular Formula: C17H18F5NO4
InChi Key: PPKHOXJANSFRFJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results