accessibility menu, dialog, popup

UserName

CAS RN 2943-75-1

H3CSiOCH3OCH3OCH3

CAS RN 2943-75-1

IUPAC Name: triethoxy(octyl)silane
Synonyms: silane, triethoxyoctyl- triethoxycaprylylsilane
Molecular Weight (g/mol): 276.49
Molecular Formula: C14H32O3Si
InChi Key: MSRJTTSHWYDFIU-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](OCC)(OCC)OCC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)