accessibility menu, dialog, popup

UserName

CAS RN 2943-70-6

IUPAC Name: (dichloromethyl)({[(dichloromethyl)dimethylsilyl]oxy})dimethylsilane
Synonyms: (dichloromethyl)({[(dichloromethyl)dimethylsilyl]oxy})dimethylsilane
Molecular Weight (g/mol): 300.15
Molecular Formula: C6H14Cl4OSi2
InChi Key: YMDCHYWOAHMASL-UHFFFAOYSA-N
SMILES: C[Si](C)(O[Si](C)(C)C(Cl)Cl)C(Cl)Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results