accessibility menu, dialog, popup

UserName

CH3FOCH3

CAS RN 29427-49-4

IUPAC Name: 1-(5-fluoro-2-methylphenyl)ethan-1-one
Synonyms: 1-(5-fluoro-2-methylphenyl)ethanone
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: KCNFQEFWFYTFAA-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results