accessibility menu, dialog, popup

UserName

KOSOOFFFFFFFFF

CAS RN 29420-49-3

IUPAC Name: potassium nonafluorobutane-1-sulfonate
Synonyms: potassium nonafluorobutane-1-sulfonate
Molecular Weight (g/mol): 338.18
Molecular Formula: C4F9KO3S
InChi Key: LVTHXRLARFLXNR-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results