CAS RN 2941-72-2
CAS RN 2941-72-2
IUPAC Name:
6-methoxy-2-methyl-1,3-benzothiazole
Synonyms:
6-methoxy-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol):
179.24
Molecular Formula:
C9H9NOS
InChi Key:
DYHLJSUORLPGNT-UHFFFAOYSA-N
SMILES:
COC1=CC=C2N=C(C)SC2=C1
1
–
1
of
1
results
1
6-Methoxy-2-methylbenzothiazole, 97%
CAS: 2941-72-2 Molecular Formula: C9H9NOS Molecular Weight (g/mol): 179.237 MDL Number: MFCD00005795 InChI Key: DYHLJSUORLPGNT-UHFFFAOYSA-N Synonym: 6-methoxy-2-methylbenzo d thiazole,6-methoxy-2-methylbenzothiazole,benzothiazole, 6-methoxy-2-methyl,2-methyl-6-methoxybenzothiazole,6-methoxy-2-methyl-benzothiazole,benzothiazole,6-methoxy-2-methyl,6-methoxy-2-methylbenzothiazol,#,benzothiazole, 6-methoxy-2-methyl-6ci,7ci,8ci,9ci PubChem CID: 76254 IUPAC Name: 6-methoxy-2-methyl-1,3-benzothiazole SMILES: CC1=NC2=C(S1)C=C(C=C2)OC