accessibility menu, dialog, popup

UserName

CAS RN 2941-62-0

H3CSNH2N

CAS RN 2941-62-0

IUPAC Name: 2-methyl-1,3-benzothiazol-6-amine
Synonyms: 2-methyl-1,3-benzothiazol-6-amine
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: HFUJOSYKJMNSFQ-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(N)C=C2S1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)