CAS RN 29394-58-9
CAS RN 29394-58-9
IUPAC Name:
1-iodo-2-methylbutane
Synonyms:
1-iodo-2-methylbutane
Molecular Weight (g/mol):
198.05
Molecular Formula:
C5H11I
InChi Key:
RHBHXHXNWHTGSO-UHFFFAOYNA-N
SMILES:
CCC(C)CI
1
–
1
of
1
results
1
(S)-(+)-1-Iodo-2-methylbutane, TRC
CAS: 29394-58-9 Molecular Formula: C5 H11 I Molecular Weight (g/mol): 198.045 Synonym: (S)-1-Iodo-2-methyl-butane,(2S)-1-Iodo-2-methylbutane,(S)-1-Iodo-2-methylbutane,(S)-2-Methylbutyl Iodide IUPAC Name: (2S)-1-iodo-2-methylbutane SMILES: CC[C@H](C)CI