accessibility menu, dialog, popup

UserName

CAS RN 29203-60-9

ClClNOHCl(E/Z)

CAS RN 29203-60-9

IUPAC Name: 2,4-dichloro-N-hydroxybenzene-1-carbonimidoyl chloride
Synonyms: 2,4-dichloro-N-hydroxybenzenecarbonimidoyl chloride
Molecular Weight (g/mol): 224.47
Molecular Formula: C7H4Cl3NO
InChi Key: GXBVZCSCOBTMHS-UHFFFAOYSA-N
SMILES: ON=C(Cl)C1=C(Cl)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results