accessibility menu, dialog, popup

UserName

CAS RN 2905-67-1

CH3OOClCl

CAS RN 2905-67-1

IUPAC Name: methyl 3,5-dichlorobenzoate
Synonyms: methyl 3,5-dichlorobenzoate
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: BTEVDFJXGLQUDS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC(Cl)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (3)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)