accessibility menu, dialog, popup

UserName

CAS RN 2905-21-7

FSClOO

CAS RN 2905-21-7

IUPAC Name: 2-fluorobenzene-1-sulfonyl chloride
Synonyms: 2-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 194.6
Molecular Formula: C6H4ClFO2S
InChi Key: ZSZKAQCISWFDCQ-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1S(Cl)(=O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)