accessibility menu, dialog, popup

UserName

CAS RN 28989-50-6

NH2SNS

CAS RN 28989-50-6

IUPAC Name: 4-(thiophen-2-yl)-1,3-thiazol-2-amine
Synonyms: 4-(thiophen-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 182.26
Molecular Formula: C7H6N2S2
InChi Key: XRNPDKQNHIKARG-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results