accessibility menu, dialog, popup

UserName

CAS RN 28920-43-6

IUPAC Name: (9H-fluoren-9-yl)methyl carbonochloridate
Synonyms: fluorenylmethyloxycarbonyl chloride
Molecular Weight (g/mol): 258.7
Molecular Formula: C15H11ClO2
InChi Key: IRXSLJNXXZKURP-UHFFFAOYSA-N
SMILES: ClC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)