accessibility menu, dialog, popup

UserName

CAS RN 289039-22-1

H3CBrCl

CAS RN 289039-22-1

IUPAC Name: 4-bromo-1-chloro-2-ethylbenzene
Synonyms: 4-bromo-1-chloro-2-ethylbenzene
Molecular Weight (g/mol): 219.51
Molecular Formula: C8H8BrCl
InChi Key: ILQNJGBEXZUYOI-UHFFFAOYSA-N
SMILES: CCC1=C(Cl)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results