accessibility menu, dialog, popup

UserName

CAS RN 28888-44-0

H3COHNONHOOCH3

CAS RN 28888-44-0

IUPAC Name: 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms: 6,7-dimethoxy-1,3-dihydroquinazoline-2,4-dione
Molecular Weight (g/mol): 222.2
Molecular Formula: C10H10N2O4
InChi Key: KWNQIIMVPSMYEM-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(=O)NC(=O)NC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results