accessibility menu, dialog, popup

UserName

CAS RN 28875-17-4

H3COO(S)CH3NHOOCH3CH3H3C

CAS RN 28875-17-4

IUPAC Name: methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: methyl (2S)-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: GJDICGOCZGRDFM-LURJTMIESA-N
SMILES: COC(=O)[C@H](C)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)