accessibility menu, dialog, popup

UserName

H3COOOH

CAS RN 288401-04-7

IUPAC Name: 2-(4-hydroxyphenyl)-6-methyl-4H-chromen-4-one
Synonyms: 2-(4-hydroxyphenyl)-6-methylchromen-4-one
Molecular Weight (g/mol): 252.27
Molecular Formula: C16H12O3
InChi Key: YAACYYNCHMHECD-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC(=CC(=O)C2=C1)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results