accessibility menu, dialog, popup

UserName

CAS RN 2882-15-7

CH3OHNH3COO

CAS RN 2882-15-7

IUPAC Name: 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate
Synonyms: 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate
Molecular Weight (g/mol): 218.23
Molecular Formula: C12H12NO3
InChi Key: TXWGINUZLBAKDF-UHFFFAOYSA-M
SMILES: COC1=CC=C2NC(C)=C(CC([O-])=O)C2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)