accessibility menu, dialog, popup

UserName

CAS RN 288-88-0

IUPAC Name: 1H-1,2,4-triazole
Synonyms: 1,2,4-triazole potassium salt 1,2,4-triazole
Molecular Weight (g/mol): 69.07
Molecular Formula: C2H3N3
InChi Key: NSPMIYGKQJPBQR-UHFFFAOYSA-N
SMILES: N1C=NC=N1
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (2)
  • (2)
Certifications/Compliance 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (7)
  • (3)
  • (1)
  • (4)
  • (1)