CAS RN 288-42-6
CAS RN 288-42-6
IUPAC Name:
1,3-oxazole
Synonyms:
oxazole
Molecular Weight (g/mol):
69.06
Molecular Formula:
C3H3NO
InChi Key:
ZCQWOFVYLHDMMC-UHFFFAOYSA-N
SMILES:
O1C=CN=C1
1
–
1
of
1
results
1
Oxazole, 98+%
CAS: 288-42-6 Molecular Formula: C3H3NO Molecular Weight (g/mol): 69.063 MDL Number: MFCD00009751 InChI Key: ZCQWOFVYLHDMMC-UHFFFAOYSA-N Synonym: oxazole,unii-fjz20i1lps,fjz20i1lps,oxazol,1,3-oxazol,pubchem8626,oxazole 1g PubChem CID: 9255 ChEBI: CHEBI:35597 IUPAC Name: 1,3-oxazole SMILES: C1=COC=N1