accessibility menu, dialog, popup

UserName

CAS RN 2878-14-0

CH3H2CH2N

CAS RN 2878-14-0

IUPAC Name: 2-methylprop-2-en-1-amine
Synonyms: 2-methylprop-2-en-1-amine
Molecular Weight (g/mol): 71.12
Molecular Formula: C4H9N
InChi Key: VXDHQYLFEYUMFY-UHFFFAOYSA-N
SMILES: CC(=C)CN
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results