accessibility menu, dialog, popup

UserName

CAS RN 2874-33-1

H3COHNOHO(E)

CAS RN 2874-33-1

IUPAC Name: (4E)-4-[(hydroxyamino)methylidene]-2-methoxycyclohexa-2,5-dien-1-one
Synonyms: (4E)-4-[(hydroxyamino)methylidene]-2-methoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: WKYQAOKXBUWYNX-AATRIKPKSA-N
SMILES: COC1=C\C(=C\NO)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results