CAS RN 28733-43-9
IUPAC Name:
5-bromopyridine-3-carboxamide
Synonyms:
5-bromonicotinamide C6H5BrN2O
Molecular Weight (g/mol):
201.02
Molecular Formula:
C6H5BrN2O
InChi Key:
YOQRXZIMSKLRCY-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CN=CC(Br)=C1
1
–
1
of
1
results
1
5-Bromonicotinamide, 98%, Thermo Scientific™
CAS: 28733-43-9 Molecular Formula: C6H5BrN2O Molecular Weight (g/mol): 201.023 MDL Number: MFCD00173919 InChI Key: YOQRXZIMSKLRCY-UHFFFAOYSA-N Synonym: 5-bromonicotinamide,5-bromo-3-pyridinecarboxamide,nicotinamide, 5-bromo,3-pyridinecarboxamide, 5-bromo,5-bromo-nicotinamide,brt,pubchem1212,acmc-1cfs0,3-bromo-5-carbamoylpyridine,5-22-02-00182 beilstein handbook reference PubChem CID: 1808 IUPAC Name: 5-bromopyridine-3-carboxamide SMILES: C1=C(C=NC=C1Br)C(=O)N