accessibility menu, dialog, popup

UserName

CAS RN 28711-29-7

OONNN

CAS RN 28711-29-7

IUPAC Name: 2-(1H-1,2,4-triazol-1-yl)acetate
Synonyms: 2-(1,2,4-triazol-1-yl)acetate
Molecular Weight (g/mol): 126.1
Molecular Formula: C4H4N3O2
InChi Key: RXDBSQXFIWBJSR-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results