accessibility menu, dialog, popup

UserName

CAS RN 28657-80-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Quantity 
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)