accessibility menu, dialog, popup

UserName

CH3CH3(S)NHO(S)CH3HNO(S)OH(S)CH3CH3NHO(S)HNO(S)NHH3COH3CCH3H3CCH3(S)OHOHO

CAS RN 28575-34-0

IUPAC Name: (3S,4S)-4-[(2S)-2-[(3S,4S)-4-[(2S)-2-[(2S)-2-acetamido-3-methylbutanamido]-3-methylbutanamido]-3-hydroxy-6-methylheptanamido]propanamido]-3-hydroxy-6-methylheptanoic acid
Synonyms: (3S,4S)-4-[(2S)-2-[(3S,4S)-4-[(2S)-2-[(2S)-2-acetamido-3-methylbutanamido]-3-methylbutanamido]-3-hydroxy-6-methylheptanamido]propanamido]-3-hydroxy-6-methylheptanoic acid
Molecular Weight (g/mol): 643.82
Molecular Formula: C31H57N5O9
InChi Key: WKYBEGDEGRCZNF-LBTYKNIQSA-N
SMILES: CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(C)=O)C(C)C)C(C)C)[C@@H](O)CC(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results