accessibility menu, dialog, popup

UserName

CH3SiCH3CH3Br

CAS RN 2857-97-8

IUPAC Name: bromotrimethylsilane
Synonyms: bromotrimethylsilane
Molecular Weight (g/mol): 153.09
Molecular Formula: C3H9BrSi
InChi Key: IYYIVELXUANFED-UHFFFAOYSA-N
SMILES: C[Si](C)(C)Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (5)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results