accessibility menu, dialog, popup

UserName

O

CAS RN 285-67-6

IUPAC Name: 6-oxabicyclo[3.1.0]hexane
Synonyms: 6-oxabicyclo 3.1.0 hexane
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: GJEZBVHHZQAEDB-UHFFFAOYNA-N
SMILES: C1CC2OC2C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results