accessibility menu, dialog, popup

UserName

H3COOOFOOH3COOCH3OOCH3

CAS RN 2823-44-1

IUPAC Name: [3,4,5-tris(acetyloxy)-6-fluorooxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-fluorooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 350.3
Molecular Formula: C14H19FO9
InChi Key: JJXATNWYELAACC-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(F)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results