accessibility menu, dialog, popup

UserName

NH2NOOOFFF

CAS RN 2822-50-6

IUPAC Name: 3-nitro-4-(trifluoromethoxy)aniline
Synonyms: 3-nitro-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 222.12
Molecular Formula: C7H5F3N2O3
InChi Key: QXIKEFNBFUHDHK-UHFFFAOYSA-N
SMILES: NC1=CC(=C(OC(F)(F)F)C=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results