CAS RN 281678-73-7
CAS RN 281678-73-7
IUPAC Name:
5-bromo-2,3-dihydro-1-benzofuran-7-carbaldehyde
Synonyms:
5-bromo-2,3-dihydro-1-benzofuran-7-carbaldehyde
Molecular Weight (g/mol):
227.06
Molecular Formula:
C9H7BrO2
InChi Key:
LQCBHNHULMHKFB-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(OCC2)C(C=O)=C1
1
–
1
of
1
results
1
5-Bromo-2,3-dihydrobenzo[b]furan-7-carbaldehyde, 97%, Thermo Scientific™
CAS: 281678-73-7 Molecular Formula: C9H7BrO2 Molecular Weight (g/mol): 227.057 InChI Key: LQCBHNHULMHKFB-UHFFFAOYSA-N Synonym: 5-bromo-2,3-dihydrobenzo b furan-7-carbaldehyde,5-bromo-2,3-dihydrobenzofuran-7-carbaldehyde,5-bromo-2,3-dihydobenzo b furan-7-aldehyde,5-bromo-2,3-dihydrobenzofuran 7-carbaldehyde,5-bromo-2,3-dihydrobenzo b furane-7-carbaldehyde,7-benzofurancarboxaldehyde, 5-bromo-2,3-dihydro PubChem CID: 2794804 IUPAC Name: 5-bromo-2,3-dihydro-1-benzofuran-7-carbaldehyde SMILES: C1COC2=C1C=C(C=C2C=O)Br