accessibility menu, dialog, popup

UserName

HOOOHN(E)

CAS RN 28166-41-8

IUPAC Name: (2E)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
Synonyms: α-cyano-4-hydroxycinnamic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: AFVLVVWMAFSXCK-VMPITWQZSA-N
SMILES: OC(=O)C(=C\C1=CC=C(O)C=C1)\C#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results