CAS RN 2812-28-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
H-L-MeThr-OH, TRC
CAS: 2812-28-4 Molecular Formula: C5H11NO3 Molecular Weight (g/mol): 133.15 Synonym: N-Methyl-L-threonine,N-Methylthreonine IUPAC Name: (2S,3R)-3-hydroxy-2-(methylamino)butanoic acid SMILES: CN[C@@H]([C@@H](C)O)C(=O)O