accessibility menu, dialog, popup

UserName

CAS RN 2809-67-8

H3CCH3

CAS RN 2809-67-8

IUPAC Name: oct-2-yne
Synonyms: 2-octyne
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: QCQALVMFTWRCFI-UHFFFAOYSA-N
SMILES: CCCCCC#CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results