accessibility menu, dialog, popup

UserName

CH3HOPOHOHOPOHOHO

CAS RN 2809-21-4

IUPAC Name: (1-hydroxy-1-phosphonoethyl)phosphonic acid
Synonyms: etidronic acid xidiphon didronel IV dequest Z 010 tetrapotassium salt etidronate ehdp, dicalcium EHDP dequest 2015 dequest 2010 ehdp - etidronate disodium
Molecular Weight (g/mol): 206.03
Molecular Formula: C2H8O7P2
InChi Key: DBVJJBKOTRCVKF-UHFFFAOYSA-N
SMILES: CC(O)(P(O)(O)=O)P(O)(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)