accessibility menu, dialog, popup

UserName

CAS RN 28036-91-1

NH2HNOClCl

CAS RN 28036-91-1

IUPAC Name: 3,4-dichlorobenzohydrazide
Synonyms: 3,4-dichlorobenzohydrazide
Molecular Weight (g/mol): 205.04
Molecular Formula: C7H6Cl2N2O
InChi Key: VEDIADQJSGNNQE-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results