accessibility menu, dialog, popup

UserName

CAS RN 2802-68-8

IUPAC Name: boron(3+) hydrogen methanol methanolate trifluoride
Synonyms: boron(3+) hydrogen methanol methoxide trifluoride
Molecular Weight (g/mol): 131.89
Molecular Formula: C2H8BF3O2
InChi Key: WWNADQBMJRRNDS-UHFFFAOYSA-L
SMILES: [H+].[B+3].[F-].[F-].[F-].CO.C[O-]
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)