accessibility menu, dialog, popup

UserName

ClAuClClClK

CAS RN 27988-75-6

IUPAC Name: potassium tetrachlorogolduide hydrate
Synonyms: potassium hydrate tetrachloroaurate
Molecular Weight (g/mol): 395.88
Molecular Formula: AuCl4H2KO
InChi Key: BGVPYGKVUIFYCE-UHFFFAOYSA-J
SMILES: O.[K+].Cl[Au-](Cl)(Cl)Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)