CAS RN 279261-89-1
CAS RN 279261-89-1
IUPAC Name:
(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)boronic acid
Synonyms:
3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid
Molecular Weight (g/mol):
193.99
Molecular Formula:
C9H11BO4
InChi Key:
CSNCBPVLUIFJOS-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C2OCCCOC2=C1
1
–
1
of
1
results
1
3,4-Dihydro-2H-1,5-benzodioxepin-7-ylboronic acid, 90+%, Thermo Scientific™
CAS: 279261-89-1 Molecular Formula: C9H11BO4 Molecular Weight (g/mol): 193.99 MDL Number: MFCD02681910 InChI Key: CSNCBPVLUIFJOS-UHFFFAOYSA-N Synonym: 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl boronic acid,3,4-dihydro-2h-1,5-benzodioxepine-7-boronic acid,3,4-dihydro-2h-1,5-benzodioxepin-7-yl boronic acid,boronic acid,b-3,4-dihydro-2h-1,5-benzodioxepin-7-yl,3,4-dihydro-2h-benzo b 1,4 dioxepin-7-ylboronic acid,boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl,pubchem7845,acmc-1crzt,3,4-dihydro-2h-1,5-benzodioxepin-7-boronic acid PubChem CID: 2776398 IUPAC Name: 3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid SMILES: OB(O)C1=CC=C2OCCCOC2=C1