CAS RN 2783-17-7
IUPAC Name:
dodecane-1,12-diamine
Synonyms:
1,12-dodecamethylenediamine
Molecular Weight (g/mol):
200.37
Molecular Formula:
C12H28N2
InChi Key:
QFTYSVGGYOXFRQ-UHFFFAOYSA-N
SMILES:
NCCCCCCCCCCCCN
1
–
1
of
1
results
1
1,12-Diaminododecane, 98+%
CAS: 2783-17-7 Molecular Formula: C12H28N2 Molecular Weight (g/mol): 200.37 MDL Number: MFCD00008155 InChI Key: QFTYSVGGYOXFRQ-UHFFFAOYSA-N Synonym: 1,12-diaminododecane,1,12-dodecanediamine,dodecamethylenediamine,dodecyldiamine,1,12-dodecamethylenediamine,1,12-diamindodecane,1,12'-dodecylenediamine,1,12'-dodecamethylenediamine,unii-j3lm80w9nt,1,12-dodecylenediamine PubChem CID: 17727 ChEBI: CHEBI:49385 IUPAC Name: dodecane-1,12-diamine SMILES: C(CCCCCCN)CCCCCN