accessibility menu, dialog, popup

UserName

CAS RN 27757-86-4

IUPAC Name: 1-(thiophen-3-yl)methanamine
Synonyms: 1-(thiophen-3-yl)methanamine
Molecular Weight (g/mol): 113.18
Molecular Formula: C5H7NS
InChi Key: DUDAKCCDHRNMDJ-UHFFFAOYSA-N
SMILES: NCC1=CSC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)