accessibility menu, dialog, popup

UserName

CAS RN 27721-02-4

PP

CAS RN 27721-02-4

IUPAC Name: [5-(diphenylphosphanyl)pentyl]diphenylphosphane
Synonyms: [5-(diphenylphosphanyl)pentyl]diphenylphosphane
Molecular Weight (g/mol): 440.51
Molecular Formula: C29H30P2
InChi Key: MZFPAWGWFDGCHP-UHFFFAOYSA-N
SMILES: C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)