accessibility menu, dialog, popup

UserName

NNSO

CAS RN 27643-15-8

IUPAC Name: 1,2,3-thiadiazole-4-carbaldehyde
Synonyms: 1,2,3-thiadiazole-4-carbaldehyde
Molecular Weight (g/mol): 114.12
Molecular Formula: C3H2N2OS
InChi Key: JNEBZFFTOLBIKJ-UHFFFAOYSA-N
SMILES: O=CC1=CSN=N1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results