accessibility menu, dialog, popup

UserName

CAS RN 2753-30-2

CH3OOH3CCH3OH3CH3COOOCH3O

CAS RN 2753-30-2

IUPAC Name: 7-(furan-3-yl)-1,8,12,16,16-pentamethyl-5,15-dioxo-3,6-dioxapentacyclo[9.8.0.0²,⁴.0²,⁸.0¹²,¹⁷]nonadec-13-en-19-yl acetate
Synonyms: 7-(furan-3-yl)-1,8,12,16,16-pentamethyl-5,15-dioxo-3,6-dioxapentacyclo[9.8.0.0²,⁴.0²,⁸.0¹²,¹⁷]nonadec-13-en-19-yl acetate
Molecular Weight (g/mol): 482.57
Molecular Formula: C28H34O7
InChi Key: YJXDGWUNRYLINJ-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC2C(C)(C)C(=O)C=CC2(C)C2CCC3(C)C(OC(=O)C4OC34C12C)C1=COC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results