accessibility menu, dialog, popup

UserName

CAS RN 275-51-4

CAS RN 275-51-4

IUPAC Name: azulene
Synonyms: azulene azunamic bicyclo(5.3.0)decapentaene azulekeep cyclopentacycloheptene
Molecular Weight (g/mol): 128.17
Molecular Formula: C10H8
InChi Key: CUFNKYGDVFVPHO-UHFFFAOYSA-N
SMILES: C1=CC2=CC=CC=CC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)