accessibility menu, dialog, popup

UserName

CAS RN 274676-47-0

H3CNNH

CAS RN 274676-47-0

IUPAC Name: 7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine
Synonyms: 2-methyl-5,6,7,8-tetrahydro-1,8-naphthyridine
Molecular Weight (g/mol): 148.21
Molecular Formula: C9H12N2
InChi Key: HBWCJMBLUCNJIS-UHFFFAOYSA-N
SMILES: CC1=NC2=C(CCCN2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results