CAS RN 27464-82-0
CAS RN 27464-82-0
IUPAC Name:
dimethyl-1,3,4-thiadiazole
Synonyms:
dimethyl-1,3,4-thiadiazole
Molecular Weight (g/mol):
114.17
Molecular Formula:
C4H6N2S
InChi Key:
JXQGICFGPUAVLJ-UHFFFAOYSA-N
SMILES:
CC1=NN=C(C)S1
1
–
1
of
1
results
1
2,5-Dimethyl-1,3,4-thiadiazole, 99+%, Thermo Scientific™
CAS: 27464-82-0 Molecular Formula: C4H6N2S Molecular Weight (g/mol): 114.166 MDL Number: MFCD00020722 InChI Key: JXQGICFGPUAVLJ-UHFFFAOYSA-N PubChem CID: 96922 IUPAC Name: 2,5-dimethyl-1,3,4-thiadiazole SMILES: CC1=NN=C(S1)C