accessibility menu, dialog, popup

UserName

CAS RN 27452-17-1

IUPAC Name: 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Synonyms: 6-bromo-1,1,4,4-tetramethyl-2,3-dihydronaphthalene
Molecular Weight (g/mol): 267.21
Molecular Formula: C14H19Br
InChi Key: NLOOVMVNNNYLFS-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=CC(Br)=CC=C12
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)