accessibility menu, dialog, popup

UserName

CAS RN 273727-50-7

IUPAC Name: 3-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
Synonyms: 3-(3-methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: YUNJEYQSCYYAEF-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC(C=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results